missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Kir2.1 Antibody, R&D Systems™

Rabbit Monoclonal Antibody
£178.00 - £446.00
Specifications
| Antigen | Kir2.1 |
|---|---|
| Clone | 2153C |
| Dilution | Western Blot 3 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 2-25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18724758
![]() |
R&D Systems
MAB9548-SP |
25 μg |
£178.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18788189
![]() |
R&D Systems
MAB9548-100 |
100 μg |
£446.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Kir2.1 Monoclonal specifically detects Kir2.1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.Specifications
| Kir2.1 | |
| Western Blot 3 ug/mL, Immunohistochemistry 3-25 ug/mL, Immunocytochemistry 2-25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human, Mouse | |
| P63252 | |
| 3759 | |
| IgG | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 2153C | |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| ATFB9, Cardiac inward rectifier potassium channel, HHBIRK1, HHIRK1, HIRK1, Inward rectifier K(+) channel Kir2.1, inward rectifier K+ channel KIR2.1, inward rectifier potassium channel 2, IRK1, IRK-1, IRK1LQT7, KCNJ2, Kir2.1, LQT7, Potassium channel, inwardly rectifying subfamily J member 2, potassium inwardly-rectifying channel, subfamily J, member 2, SQT3 | |
| E. coli-derived recombinant human Kir2.1 Asn352-Ile427 Accession # P63252 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Kir2.1 in direct ELISAs. Detects human and mouse Kir2.1 in Immunohistochemistry. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title