missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse JunD Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5526-SP
This item is not returnable.
View return policy
Description
JunD Polyclonal specifically detects JunD in Human, Mouse samples. It is validated for Western Blot.
Specifications
| JunD | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| activator protein 1, AP-1, jun D proto-oncogene, JunD-FL isoform, transcription factor jun-D | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3727 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P17535 | |
| JUND | |
| E. coli-derived recombinant human JunD Lys47-Gly156 Accession # P17535 | |
| 25 μg | |
| Primary | |
| Detects human and mouse JunD in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction