missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Jumonji/JARID2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6090-SP
This item is not returnable.
View return policy
Description
Jumonji/JARID2 Polyclonal specifically detects Jumonji/JARID2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Jumonji/JARID2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| JARID2, JMJ, JMJjumonji-like protein, jumonji (mouse) homolog, jumonji homolog, jumonji, AT rich interactive domain 2, Jumonji/ARID domain-containing protein 2, protein Jumonji | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 3720 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q92833 | |
| JARID2 | |
| E. coli-derived recombinant human Jumonji/JARID2 Met1-Thr159 Accession # Q92833 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Jumonji/JARID2 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human JARID2B is observed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction