missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Integrin alpha 11 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB4235-SP
This item is not returnable.
View return policy
Description
Integrin alpha 11 Monoclonal specifically detects Integrin alpha 11 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Integrin alpha 11 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| EAW77820 | |
| ITGA11 | |
| Chinese hamster ovary cell line CHO-derived recombinant human Integrin alpha 11 Phe23-Pro1142 (Leu972Pro, Val1030 del) Accession # EAW77820 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Integrin alpha 11 in direct ELISAs and Western blots. In direct ELISAs, does not cross-react with recombinant human Integrin alpha 2, alpha 10, alpha X, or alpha L. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 396214 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HsT18964, integrin alpha-11, integrin, alpha 11, ITGA11 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 22801 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction