missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse IL-12 R beta 2 APC-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 4 publications
£254.00 - £461.00
Specifications
| Antigen | IL-12 R beta 2 |
|---|---|
| Clone | 305719 |
| Dilution | Flow Cytometry 10 uL/10^6 cells |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18759108
![]() |
R&D Systems
FAB1959A-025 |
25 Tests |
£254.00
25 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18718917
![]() |
R&D Systems
FAB1959A-100 |
100 Tests |
£461.00
100 tests |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-12 R beta 2 Monoclonal specifically detects IL-12 R beta 2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
| IL-12 R beta 2 | |
| Flow Cytometry 10 uL/10^6 cells | |
| Monoclonal | |
| Purified | |
| RUO | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| IL-12 receptor beta 2, IL-12 receptor subunit beta-2, IL12R beta 2, IL-12R subunit beta-2, IL12RB2, IL-12RB2, IL-12R-beta-2, interleukin 12 receptor, beta 2, interleukin-12 receptor beta-2 chain, interleukin-12 receptor subunit beta-2 | |
| IL12RB2 | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Detects human and mouse IL-12 R beta 2 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant human IL-1 R beta 1 is observed. |
| 305719 | |
| Flow Cytometry | |
| APC | |
| Mouse | |
| Human, Mouse | |
| Q99665 | |
| 3595 | |
| Mouse myeloma cell line NS0-derived recombinant human IL-12 R beta 2 Cys28-Asn622 Accession # Q99665 | |
| Primary | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title