missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse HSP10/EPF Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3298-SP
This item is not returnable.
View return policy
Description
HSP10/EPF Polyclonal specifically detects HSP10/EPF in Human, Mouse samples. It is validated for Western Blot.
Specifications
| HSP10/EPF | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 10 kDa chaperonin, 10 kDa heat shock protein, mitochondrial, Chaperonin 10, Chaperonin 10 Homolog, CPN10HSP10, Early-pregnancy factor, EPF, GroES, heat shock 10kD protein 1 (chaperonin 10), heat shock 10kDa protein 1 (chaperonin 10), Hsp10, HSPE1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3336 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P61604 | |
| HSPE1 | |
| E. coli-derived recombinant human HSP10/EPF Ala2-Asp102 Accession # P61604 | |
| 25 μg | |
| Primary | |
| Detects endogenous human and mouse HSP10/EPF in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction