missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6085-SP
This item is not returnable.
View return policy
Description
Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 Polyclonal specifically detects Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Heparan Sulfate 3-O-Sulfotransferase 4/HS3ST4 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 30ST4, 3-OST-4, 3OST4 heparan sulfate 3-O-sulfotransferase-4, EC 2.8.2, EC 2.8.2.23, h3-OST-4, heparan sulfate (glucosamine) 3-O-sulfotransferase 4, Heparan sulfate 3-O-sulfotransferase 4, Heparan sulfate D-glucosaminyl 3-O-sulfotransferase 4, heparan sulfate glucosamine 3-O-sulfotransferase 4 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 9951 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Neutralization | |
| Q9Y661 | |
| HS3ST4 | |
| E. coli-derived recombinant human Heparan Sulfate 3-O-Sulfotransferase4/HS3ST4 Gly184-Lys456 Accession # Q9Y661 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Heparan Sulfate 3-O-Sulfotransferase4/HS3ST4 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction