missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Golgi Glycoprotein 1/GLG1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB78791
This item is not returnable.
View return policy
Description
Golgi Glycoprotein 1/GLG1 Monoclonal specifically detects Golgi Glycoprotein 1/GLG1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Golgi Glycoprotein 1/GLG1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CFR1, CFR-1, cysteine-rich fibroblast growth factor receptor, DKFZp686L08213, E-selectin ligand 1, ESL1, ESL-1, FLJ23319, FLJ23967, FLJ32797, GLG1, Golgi apparatus protein 1, golgi glycoprotein 1, golgi sialoglycoprotein MG-160, MG160, MG-160 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2734 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 858238 | |
| Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q92896 | |
| GLG1 | |
| E. coli-derived recombinant human Golgi Glycoprotein 1/GLG1 Lys1048-Asn1145 Accession # Q92896 | |
| 100 μg | |
| Primary | |
| Detects human Golgi Glycoprotein 1/GLG1 in direct ELISA. Detects human and mouse GLG1 in Western Blots. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction