missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Glucokinase/GCK Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB7840
This item is not returnable.
View return policy
Description
Glucokinase/GCK Monoclonal specifically detects Glucokinase/GCK in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Glucokinase/GCK | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P35557 | |
| GCK | |
| E. coli-derived recombinant human Glucokinase/GCK Val16-Gln465 Accession # P35557 | |
| 100 μg | |
| Primary | |
| Detects human Glucokinase/GCK in ELISAs. In direct ELISAs, no cross-reactivity with recombinant human PRF-1 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 849520 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 2.7.1, EC 2.7.1.2, FGQTL3, GCK, GK, GLK, glucokinase (hexokinase 4), hexokinase-4, Hexokinase-D, HHF3, HK4, HK4pancreatic isozyme, HKIV, HXKP, LGLK, maturity onset diabetes of the young 2, MODY2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2645 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction