missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse GATA-2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF2046
This item is not returnable.
View return policy
Description
GATA-2 Polyclonal specifically detects GATA-2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| GATA-2 | |
| Polyclonal | |
| Unconjugated | |
| P23769 | |
| GATA2 | |
| E. coli-derived recombinant human GATA-2 Ala15-Thr279 Accession # P23769 | |
| 100 μg | |
| Primary | |
| Detects human and mouse GATA-2 in Western blots. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| endothelial transcription factor GATA-2, FLJ45948, GATA binding protein 2, GATA2, GATA-binding protein 2NFE1B, MGC2306 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2624 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction