missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Frizzled-7 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 4 publications
£155.00 - £387.00
Specifications
| Antigen | Frizzled-7 |
|---|---|
| Clone | 151143 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunocytochemistry, CyTOF |
Description
Frizzled-7 Monoclonal specifically detects Frizzled-7 in Human, Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry, CyTOF-ready.Specifications
| Frizzled-7 | |
| LYOPH | |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| Q61090 | |
| 8324 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-7 Gln33-Leu185 Accession # Q61090 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse Frizzled-7. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -3, -4 or -8 is observed. |
| 151143 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| frizzled (Drosophila) homolog 7, frizzled homolog 7 (Drosophila), Frizzled, drosophila, homolog of, 7, Frizzled7, fz-7, FZD7, FzE3frizzled-7, hFz7 | |
| FZD7 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title