missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Frizzled-4 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB194-SP
This item is not returnable.
View return policy
Description
Frizzled-4 Monoclonal specifically detects Frizzled-4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Frizzled-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q61088 | |
| FZD4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-4 Phe37-Glu180 Accession # Q61088 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Frizzled-4 in Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -6, -7, -8, or recombinant human Frizzled-5 is observed. Detects Frizzled-4 by flow cytometry and immunohistochemistry on mouse differentiated embryoid bodies derived from the mouse embryonic cell line, D3. | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 145901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD344, CD344 antigen, EVR1, exudative vitreoretinopathy 1, FEVR, frizzled (Drosophila) homolog 4, frizzled homolog 4 (Drosophila), frizzled-4, Frizzled4, Fz-4, FZD4, FZD4S, FzE4, GPCR, hFz4, MGC34390, WNT receptor frizzled-4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8322 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction