missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Frizzled-4 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB194-050
This item is not returnable.
View return policy
Description
Frizzled-4 Monoclonal specifically detects Frizzled-4 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
| Frizzled-4 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
| Q61088 | |
| FZD4 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-4 Phe37-Glu180 Accession # Q61088 | |
| 50 μg | |
| Primary | |
| Detects human and mouse Frizzled-4 in Western blots. In Western blots, no cross-reactivity with recombinant mouse (rm) Frizzled-1, -2, -3, -6, -7, -8, or recombinant human Frizzled-5 is observed. Detects Frizzled-4 by flow cytometry and immunohistochemistry on mouse differentiated embryoid bodies derived from the mouse embryonic cell line, D3. | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF | |
| 145901 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD344, CD344 antigen, EVR1, exudative vitreoretinopathy 1, FEVR, frizzled (Drosophila) homolog 4, frizzled homolog 4 (Drosophila), frizzled-4, Frizzled4, Fz-4, FZD4, FZD4S, FzE4, GPCR, hFz4, MGC34390, WNT receptor frizzled-4 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8322 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion