missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Frizzled-3 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
£167.00 - £379.00
Specifications
| Antigen | Frizzled-3 |
|---|---|
| Clone | 169310 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Immunohistochemistry, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18794377
![]() |
R&D Systems
MAB1001-SP |
25 μg |
£167.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18172211
|
R&D Systems
MAB1001-050 |
50 μg |
£258.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18173091
|
R&D Systems
MAB1001-100 |
100 μg |
£379.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Frizzled-3 Monoclonal specifically detects Frizzled-3 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready.Specifications
| Frizzled-3 | |
| LYOPH | |
| Flow Cytometry, Immunohistochemistry, CyTOF | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| Q61086 | |
| 7976 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-3 Met1-Val157 Accession # Q61086 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects mouse Frizzled-3 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse Frizzled-1, -4, -7, or -8 is observed. By flow cytometry and immunohistochemistry, this antibody also detects mouse Frizzled-3 on differentiated embryoid bodies derived from the mouse embryonic cell line, D3. This antibody also detects Frizzled-3 on human HEK293 cells by immunocytochemistry and flow cytometry. |
| 169310 | |
| Flow Cytometry 2.5 ug/10^6 cells, Immunohistochemistry 8-25 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| frizzled (Drosophila) homolog 3, frizzled homolog 3 (Drosophila), frizzled-3, Frizzled3, Fz-3, FZD3, hFz3 | |
| FZD3 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title