missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Frizzled-2 Antibody, R&D Systems™
Rat Monoclonal Antibody has been used in 1 publication
Brand: R&D Systems MAB1307-050
This item is not returnable.
View return policy
Description
Frizzled-2 Monoclonal specifically detects Frizzled-2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Frizzled-2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation | |
| Q9JIP6 | |
| FZD2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Frizzled-2 Gln29-Leu168 Accession # Q9JIP6 | |
| 50 μg | |
| Primary | |
| Detects mouse Frizzled-2 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody does not cross-react with recombinant mouse (rm) Frizzled-1, -3, -4, -6, -7, -8, or rhFrizzled-5. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 202102 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| frizzled (Drosophila) homolog 2, frizzled homolog 2 (Drosophila), frizzled-2, Frizzled2, fz-2, Fz2, FZD2, FzE2, hFz2 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2535 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction