missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse FoxF1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 7 publications
Brand: R&D Systems AF4798-SP
This item is not returnable.
View return policy
Description
FoxF1 Polyclonal specifically detects FoxF1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| FoxF1 | |
| Polyclonal | |
| Western Blot 1 ug/mL | |
| Q12946 | |
| FOXF1 | |
| E. coli-derived recombinant human FoxF1 Met154-Met379 Accession # Q12946 | |
| 25 μg | |
| Primary | |
| Detects human and mouse FoxF1 in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FKHL5, FKHL5MGC105125, forkhead box F1, forkhead box protein F1, Forkhead, drosophila, homolog-like 5, Forkhead-related activator 1, Forkhead-related protein FKHL5, Forkhead-related transcription factor 1, FREAC-1, FREAC1ACDMPV | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2294 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
For Research Use Only
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto