missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human/Mouse FGF-8b, Animal-Free Recombinant Protein, PeproTech®

Product Code. 17833633
Click to view available options
Quantity:
100 μg
2 x 500 μg
25 μg
500 μg
Unit Size:
100µg
1mg
25µg
500µg
Pack of 2
This item is not returnable. View return policy

Product Code. 17833633

Brand: Gibco™ AF1002525UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

AF-100-25-1MG will be provided as 2 x 500 μg (AF-100-25-500UG). Recombinant Human/Murine FGF-8 (FGF-8b) is a 22.5 kDa protein consisting of 194 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF-8 (FGF-8b) is a heparin-binding growth factor belonging to the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. There are 4 known alternate spliced forms of FGF8; FGF-8A, FGF-8B, FGF-8E and FGF-8F. The human and murine FGF-8A and B are identical, unlike human and mouse FGF-8E and F, which are 98% identical. FGF-8 targets mammary carcinoma cells and other cells expressing the FGF receptors.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P37237, P55075
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 14179, 2253
Molecular Weight (g/mol) 22.5 kDa
Name Human/Mouse FGF-8 b, Animal-Free
Quantity 25 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Fibroblast Growth Factor 8
Common Name FGF-8 B
Gene Symbol FGF8
Biological Activity Determined by a cell proliferation assay using balb/c 3T3 cells. The expected ED50 for this effect is 2.0-4.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence MQVTVQSSPN FTQHVREQSL VTDQLSRRLI RTYQLYSRTS GKHVQVLANK RINAMAEDGD PFAKLIVETD TFGSRVRVRG AETGLYICMN KKGKLIAKSN GKGKDCVFTE IVLENNYTAL QNAKYEGWYM AFTRKGRPRK GSKTRQHQRE VHFMKRLPRG HHTTEQSLRF EFLNYPPFTR SLRGSQRTWA PEPR
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 97% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.