missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse EphB2 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 35 publications
Brand: R&D Systems AF467
This item is not returnable.
View return policy
Description
EphB2 Polyclonal specifically detects EphB2 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.
Specifications
| EphB2 | |
| Polyclonal | |
| Unconjugated | |
| P54763 | |
| EPHB2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse EphB2 Val27-Lys548 Accession # P54763 | |
| 100 μg | |
| Primary | |
| Detects mouse and human EphB2 in direct ELISAs and Western blots. In Western blots, approximately 5% cross-reactivity with recombinant rat (rr) EphB1, recombinant mouse (rm) EphA8, rmEphA6, rmEphB6, rmEphA3, rmEphA4, rmEphA7, rrEphA5, recombinant human EphA1, rmEphA2 and rmEphB3 is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 20 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL, CyTOF-ready | |
| CAPB, Cek5, Drt, DRTEphB2, EC 2.7.10, EC 2.7.10.1, EK5, elk-related tyrosine kinase, EPH receptor B2, eph tyrosine kinase 3, EPH-like kinase 5, ephrin type-B receptor 2, EPHT3MGC87492, EPTH3, Erk, ERKHek5, HEK5, Nuk, PCBC, protein-tyrosine kinase HEK5, Qek2, Renal carcinoma antigen NY-REN-47, Sek3, Tyro5, Tyrosine-protein kinase receptor EPH-3, Tyrosine-protein kinase TYRO5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2048 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction