missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse EMX2 Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF6470
This item is not returnable.
View return policy
Description
EMX2 Polyclonal specifically detects EMX2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| EMX2 | |
| Polyclonal | |
| Unconjugated | |
| Q04743 | |
| EMX2 | |
| E. coli-derived recombinant human EMX2 Asn49-Ser143 Accession # Q04743 | |
| 100 μg | |
| Primary | |
| Detects human EMX2 in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| empty spiracles homeobox 2, Empty spiracles homolog 2, empty spiracles homolog 2 (Drosophila), Empty spiracles-like protein 2, homeobox protein EMX2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 2018 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction