missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse ELF3 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF5787
This item is not returnable.
View return policy
Description
ELF3/ESE-1 Polyclonal specifically detects ELF3/ESE-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry.
Specifications
| ELF3/ESE-1 | |
| Polyclonal | |
| Unconjugated | |
| E74-like factor 3, E74-like factor 3 (ets domain transcription factor, epithelial-specific ), Epithelial-restricted with serine box, epithelial-specific), Epithelium-restricted Ets protein ESX, Epithelium-specific Ets transcription factor 1, EPR1, EPR-1, ERT, ESX, ets domain transcription factor, serine box (epithelial-specific), ETS-related transcription factor Elf-3, JEN, serine box | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1999 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| ELF3 | |
| E. coli-derived recombinant human ELF3 | |
| 100 μg | |
| Primary | |
| Detects human and mouse ELF3. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction