missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse EBF-2 Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF7006-SP
This item is not returnable.
View return policy
Description
EBF-2 Polyclonal specifically detects EBF-2 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| EBF-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| COE2, EBF2, EBF-2, Mmot1, O/E-3, OE-3 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 64641 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| O08792 | |
| EBF2 | |
| E. coli-derived recombinant mouse EBF-2 Arg407-Ser519 Accession # O08792 | |
| 25 μg | |
| Primary | |
| Detects mouse EBF-2 in direct ELISAs. Detects mouse and human EBF-2 in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse (rm) EBF-1 and rmEBF-3 is observed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction