missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse DR3/TNFRSF25 Alexa Fluor 750-conjugated Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems FAB943S-100UG
This item is not returnable.
View return policy
Description
DR3/TNFRSF25 Monoclonal specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| DR3/TNFRSF25 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q93038 | |
| TNFRSF25 | |
| Mouse myeloma cell line NS0-derived recombinant human DR3 isoform 1 Gln25-Phe201 Accession # Q93038 | |
| RUO | |
| 8718 | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| 59204 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) DR3/TNFRSF25 and no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR6, rhBAFF R, rhFas, rhGITR, rhLTR beta , rhNGF R, rhOPG, rmOX40, rhRANK, rhTAJ, rhTNF RI, rhTNF RII, or rhHVEM. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction