missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Dkk-1 Antibody, R&D Systems™
Description
Produced in goats immunized with purified, insect cell line Sf 21-derived, recombinant human Dickkopf homolog-1 (rhDkk-1). Human Dkk-1 specific IgG was purified by human Dkk-1 affinity chromatography.
Dkk-1 Polyclonal specifically detects Dkk-1 in Human, Mouse samples. It is validated for Western Blot, Blockade of Receptor-ligand Interaction, Knockout Validated.
Specifications
Specifications
| Antigen | Dkk-1 |
| Applications | Western Blot, Blockade of Receptor-ligand Interaction, Immunoassay |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Blockade of Receptor-ligand Interaction, Knockout Validated |
| Gene Accession No. | O94907 |
| Gene Alias | dickkopf (Xenopus laevis) homolog 1, dickkopf homolog 1 (Xenopus laevis), dickkopf related protein-1, Dickkopf-1, dickkopf-related protein 1, Dkk1, DKK-1, hDkk-1, SKdickkopf-1 like |
| Gene Symbols | DKK1 |
| Host Species | Goat |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?