missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse DGK-iota Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6435
This item is not returnable.
View return policy
Description
DGK-iota Monoclonal specifically detects DGK-iota in Human, Mouse samples. It is validated for Western Blot.
Specifications
| DGK-iota | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O75912 | |
| DGKI | |
| E. coli-derived recombinant human DGK-iota Ser925-Val1065 (predicted) Accession # O75912 | |
| 100 μg | |
| Primary | |
| Detects human and mouse DGK-iota in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human DGK-alpha , -beta , -delta , -epsilon , -eta , -gamma , -kappa , -theta , or -zeta is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 648538 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DAG kinase iota, DGKI, DGKiota, DGK-IOTA, diacylglycerol kinase iota, diacylglycerol kinase, iota, Diglyceride kinase iota, EC 2.7.1.107 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 9162 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction