missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse DAPP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7024-SP
This item is not returnable.
View return policy
Description
DAPP1 Monoclonal specifically detects DAPP1 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| DAPP1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UN19 | |
| DAPP1 | |
| E. coli-derived recombinant human DAPP1 Gly2-Ser163 Accession # Q9UN19 | |
| 25 μg | |
| Primary | |
| Detects human DAPP1 in direct ELISAs. Detects human and mouse DAPP1 in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 716804 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| B lymphocyte adapter protein Bam32, BAM32, BAM32DKFZp667E0716, B-cell adapter molecule of 32 kDa, dual adapter for phosphotyrosine and 3-phosphotyrosine and 3-phosphoinositide, dual adaptor of phosphotyrosine and 3-phosphoinositides, hDAPP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 27071 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction