missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CXCL12/SDF-1 Antibody, R&D Systems™

Description
CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Mouse samples. It is validated for Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready, Dual RNAscope ISH-IHC, ELISA Capture (Matched Antibody Pair), ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
| Antigen | CXCL12/SDF-1 |
| Applications | Immunohistochemistry, Flow Cytometry, Immunocytochemistry, CyTOF, In Situ Hybridization (ISH), ELISA |
| Classification | Monoclonal |
| Clone | 79018 |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-15 ug/mL, CyTOF-ready, Dual RNAscope ISH-IHC 5-25 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q6ICW0 |
| Gene Alias | chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSF, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF1, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 |
| Gene Symbols | CXCL12 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?