missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD45R / B220 Rat, PE-Cy5, Clone: RA3-6B2, enQuireBio™
Rat Monoclonal Antibody
£95.92 - £204.75
Specifications
| Antigen | Human/Mouse CD45R / B220 |
|---|---|
| Clone | RA3-6B2 |
| Concentration | 0.2mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15931998
|
EnquireBio
QAB41-PE5-25ug |
25 μg |
£95.92
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15921998
|
EnquireBio
QAB41-PE5-100ug |
100 μg |
£204.75
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| Human/Mouse CD45R / B220 | |
| 0.2mg/mL | |
| Monoclonal | |
| Rat | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
| 19264 | |
| IgG2a κ | |
| Protein A/G Purified |
| RA3-6B2 | |
| Flow Cytometry | |
| PE-Cyanine5 | |
| RUO | |
| NP_001255215 | |
| Ptprc | |
| Primary | |
| Store at 2-4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title