Learn More
Human/Mouse CD44 Rat, Biotin, Clone: IM7, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB39-B-25ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 15941928
  • £62.45 / 25µg
Estimated Shipment: 19-06-2024
to see stock.
Specifications
Human/Mouse CD44
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1, pH7.2
Cd44
Protein A/G Purified
RUO
12505
IgG2b κ
Flow Cytometry
IM7
Biotin
NP_001034239
Rat
25 μg
Primary
Store at 2-4°C.