missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD44 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB6127-SP
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human, Mouse samples. It is validated for Flow Cytometry, CyTOF-reported.
Specifications
| CD44 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD44 | |
| Myeloid leukemia M1 cells induced with Dexamethasone | |
| 25 μg | |
| Primary | |
| Detects human and mouse CD44 in flow cytometry. | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, CyTOF | |
| IM7.8.1R | |
| Flow Cytometry 0.25 ug/10^6 cells, CyTOF-reported | |
| CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| 960 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction