missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse CD44 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems FAB6127G-100
This item is not returnable.
View return policy
Description
CD44 Monoclonal specifically detects CD44 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| CD44 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| CD44 | |
| Myeloid leukemia M1 cells induced with Dexamethasone | |
| 100 Tests | |
| 960.0 | |
| Human, Mouse | |
| Supernatant |
| Flow Cytometry | |
| IM7.8.1R | |
| Flow Cytometry 5 μL/106 cells | |
| CD44, CD44 antigen, CD44 molecule (Indian blood group), CD44R, CDw44, cell surface glycoprotein CD44, chondroitin sulfate proteoglycan 8, CSPG8, ECMR-III, Epican, Extracellular matrix receptor III, GP90 lymphocyte homing/adhesion receptor, HCAM, HCELL, hematopoietic cell E- and L-selectin ligand, Heparan sulfate proteoglycan, Hermes antigen, homing function and Indian blood group system, HUTCH-I, Hyaluronate receptor, IN, LHR, MC56, MDU2, MDU2CD44 antigen (homing function and Indian blood group system), MDU3, MDU3CDW44, MIC4, MIC4MGC10468, MUTCH-I, Pgp1, PGP-1, PGP-I, Phagocytic glycoprotein 1, Phagocytic glycoprotein I | |
| Rat | |
| Protein A or G purified from cell culture supernatant | |
| Primary | |
| Detects human and mouse CD44 in flow cytometry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction