missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Cathepsin B Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
£181.00 - £487.00
Specifications
| Antigen | Cathepsin B |
|---|---|
| Clone | 173317 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18729707
![]() |
R&D Systems
MAB965-SP |
25 μg |
£181.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18171101
![]() |
R&D Systems
MAB965 |
500 μg |
£487.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cathepsin B Monoclonal specifically detects Cathepsin B in Human, Mouse samples. It is validated for Western Blot.Specifications
| Cathepsin B | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, CTSB, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
| CTSB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 173317 | |
| Western Blot | |
| Unconjugated | |
| Rat | |
| Human, Mouse | |
| P10605 | |
| 1508 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin B His18-Phe339 Accession # P10605 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects the pro and active forms of human and mouse Cathepsin B in direct ELISAs and Western blots. Does not cross-react with recombinant human Cathepsin C, L, L2, O, S, Z, recombinant mouse Cathepsin C, H, L, or Z. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title