missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Caspase-8 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 5 publications
Brand: R&D Systems AF705
This item is not returnable.
View return policy
Description
Caspase-8 Polyclonal specifically detects Caspase-8 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Knockout Validated.
Specifications
| Caspase-8 | |
| Polyclonal | |
| Unconjugated | |
| AAC50645 | |
| CASP8 | |
| E. coli-derived recombinant human Caspase-8 Ser234-Asp496 Accession # AAC50645 | |
| 100 μg | |
| Primary | |
| Detects human and mouse Caspase-8 in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, KnockDown | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Knockout Validated | |
| AIS, androgen receptor, CASP8, Caspase8, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, Mch5, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 841 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction