missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Cadherin-13 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB3264-SP
This item is not returnable.
View return policy
Description
Cadherin-13 Monoclonal specifically detects Cadherin-13 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.
Specifications
| Cadherin-13 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| P55290 | |
| CDH13 | |
| Mouse myeloma cell line NS0-derived recombinant human Cadherin-13 Glu23-Ala692 Accession # P55290 | |
| 25 μg | |
| Primary | |
| Detects human Cadherin-13 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) E-Cadherin, rhN-Cadherin, rhP-Cadherin, rhVE-Cadherin, rhCadherin-8, -11, -12 or -17 is observed. | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 392411 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cadherin 13, H-cadherin (heart), cadherin-13, Cadherin13, CDH13, CDHHT-cadherin, H-cadherin, Heart cadherin, heart-cadherin, P105, T-cad, T-Cadherin, Truncated cadherin, truncated-cadherin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1012 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction