missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse c-Jun Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB2670-SP
This item is not returnable.
View return policy
Description
c-jun Monoclonal specifically detects c-jun in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| c-jun | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| P05412 | |
| JUN | |
| E. coli-derived recombinant human c-Jun Met1-Phe331 Accession # P05412 | |
| 25 μg | |
| Primary | |
| Detects human and mouse c-Jun in Western blots. | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 320831 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Activator protein 1, AP1, AP-1, cJun, c-Jun, enhancer-binding protein AP1, JUN, Jun activation domain binding protein, jun oncogene, jun proto-oncogene, Proto-oncogene c-Jun, transcription factor AP-1, V-jun avian sarcoma virus 17 oncogene homologp39, v-jun sarcoma virus 17 oncogene homolog, v-jun sarcoma virus 17 oncogene homolog (avian) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3725 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction