missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Bub3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5706-SP
This item is not returnable.
View return policy
Description
Bub3 Polyclonal specifically detects Bub3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Bub3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BUB3 (budding uninhibited by benzimidazoles 3, yeast) homolog, BUB3 budding uninhibited by benzimidazoles 3 homolog, BUB3L, budding uninhibited by benomyl, budding uninhibited by benzimidazoles 3 homolog (yeast), hBUB3, mitotic checkpoint component, mitotic checkpoint protein BUB3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9184 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O43684 | |
| BUB3 | |
| E. coli-derived recombinant human Bub3 Met1-Thr328 Accession # O43684 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Bub3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Bub1 is observed. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction