missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BID Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 6 publications
Brand: R&D Systems MAB860
This item is not returnable.
View return policy
Description
BID Monoclonal specifically detects BID in Human, Mouse samples. It is validated for Western Blot.
Specifications
| BID | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P70444 | |
| BID | |
| E. coli-derived recombinant mouse BID Met1-Asp195 Accession # P70444 | |
| 100 μg | |
| Primary | |
| Detects mouse BID in Western blots. Detects full-length mouse BID and the 15 kDa carboxyl-terminal fragment (tBID) generated by cleavage with Caspase-8. The antibody also detects human BID. However, in Western blots of recombinant human and mouse BID, the antibody is most sensitive for mouse BID. | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| 91508 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 637 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction