missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Bcl-w Antibody, R&D Systems™

Rabbit Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF824-SP
This item is not returnable.
View return policy
Description
BCL-W/BCL2L2 Polyclonal specifically detects BCL-W/BCL2L2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| BCL-W/BCL2L2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2L2, BCL2-like 2, Bclw, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 599 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1.2 ug/mL | |
| Q92843 | |
| BCL2L2 | |
| KLH-coupled human/mouse Bcl-w synthetic peptide Gly32-Asp51 Accession # Q92843 | |
| 25 μg | |
| Primary | |
| Detects human and mouse Bcl-w in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction