missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BACE-1 Ectodomain Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF931
Additional Details : Weight : 0.00970kg
Description
BACE-1 Polyclonal specifically detects BACE-1 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
BACE-1 | |
Polyclonal | |
Unconjugated | |
P56817 | |
BACE1 | |
Mouse myeloma cell line NS0-derived recombinant human BACE-1 Ectodomain Thr22-Tyr460 Accession # P56817 | |
100 μg | |
Primary | |
Detects human and mouse BACE-1 Ectodomain in direct ELISAs and Western blots. In direct ELISAs, less than 2% cross-reactivity with recombinant human (rh) ADAM8, recombinant mouse ADAM9, rhADAM15, and rhTACE is observed. | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
APP beta-secretase, ASP2, Aspartyl protease 2, BACE1, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
Goat | |
Affinity Purified | |
RUO | |
23621 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only