missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse BACE-1 Ectodomain Alexa Fluor 594-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC931T-100UG
This item is not returnable.
View return policy
Description
BACE-1 Monoclonal specifically detects BACE-1 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| BACE-1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P56817 | |
| BACE1 | |
| Mouse myeloma cell line NS0-derived recombinant human BACE-1 Thr22-Tyr460 Accession # P56817 | |
| RUO | |
| 23621 | |
| Human, Mouse | |
| IgG1 |
| Flow Cytometry | |
| 137612 | |
| Alexa Fluor 594 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| APP beta-secretase, ASP2, Aspartyl protease 2, BACE1, BACEAsp 2, beta-secretase 1, beta-secretase 1 precursor variant 1, beta-site amyloid beta A4 precursor protein-cleaving enzyme, Beta-site amyloid precursor protein cleaving enzyme 1, Beta-site APP cleaving enzyme 1, beta-site APP-cleaving enzyme, beta-site APP-cleaving enzyme 1, EC 3.4.23, EC 3.4.23.46, FLJ90568, HSPC104, KIAA1149, Memapsin-2, Membrane-associated aspartic protease 2, transmembrane aspartic proteinase Asp2 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human and mouse BACE-1 in direct ELISAs and human BACE-1 in Western blots. In Western blots, no cross-reactivity with recombinant human (rh) BACE-2, recombinant mouse (rm) BACE-2, rhADAM8, rmADAM9, rmADAM10, rhADAM15, or rhTACE is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction