Learn More
Human/Mouse Active Caspase-3 Antibody, R&D Systems™
Rabbit Polyclonal Antibody has been used in 155 publications
Brand:  R&D Systems AF835
Product Code. 18011652
  • £418.00 / 50µg
Description
Caspase-3 Polyclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Caspase-3
Polyclonal
Unconjugated
P42574
CASP3
KLH-coupled human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # P42574
50 μg
Primary
Distinguishes apoptotic cells from non-apoptotic cells in immunohistochemistry. Detects the p17 subunit of Caspase-3, but does not detect the precursor form in Western blot. We recommend R&D Systems Affinity Purified Anti-Caspase-3 (Catalog # AF-605-NA) or Monoclonal Anti-Cleaved-Caspase-3 (Catalog # MAB835) for Western blot applications.
Human, Mouse
IgG
Immunohistochemistry, Immunocytochemistry
LYOPH
Immunohistochemistry 1-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama
Rabbit
Affinity Purified
RUO
836
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.