missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse 14-3-3 zeta Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 2 publications
Brand: R&D Systems AF2669
This item is not returnable.
View return policy
Description
14-3-3 zeta Polyclonal specifically detects 14-3-3 zeta in Human, Mouse samples. It is validated for Western Blot.
Specifications
| 14-3-3 zeta | |
| Polyclonal | |
| Unconjugated | |
| P63104 | |
| YWHAZ | |
| E. coli-derived recombinant human 14-3-3 zeta Met1-Asn245 Accession # P63104 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat 14-3-3 zeta in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human 14-3-3 beta, epsilon, eta, gamma, or sigma is observed. | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.25 ug/mL | |
| 1433 zeta, KCIP-1, KCIP-1MGC126532,14-3-3-zeta, MGC111427, MGC138156, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, deltapolypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, zetapolypeptide, YWHAZ, zeta polypeptide | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7534 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction