missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human MORC2 (aa 888-1026) Control Fragment Recombinant Protein
Click to view available options
Quantity:
100 μL
Tamaño de la unidad:
100µL
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (92%), Rat (92%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-51390 (PA5-51390. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
Especificaciones
Especificaciones
| Accession Number | Q9Y6X9 |
| Concentration | ≥5.0 mg/mL |
| For Use With (Application) | Blocking Assay, Control |
| Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
| Gene ID (Entrez) | 22880 |
| Name | Human MORC2 (aa 888-1026) Control Fragment |
| Quantity | 100 μL |
| Regulatory Status | RUO |
| Gene Alias | 8430403M08Rik; ATPase MORC2; ATPase MORC2A; CMT2Z; Kiaa0852; microrchidia 2 A; MORC family CW-type zinc finger 2; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2 A; MORC2; Morc2a; ZCW3; ZCWCC1; zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1 |
| Common Name | MORC2 |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido