missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMR/CD206 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 3 publications
Brand: R&D Systems MAB25341-SP
This item is not returnable.
View return policy
Description
MMR/CD206/Mannose Receptor Monoclonal specifically detects MMR/CD206/Mannose Receptor in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| MMR/CD206/Mannose Receptor | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P22897 | |
| MRC1 | |
| Mouse myeloma cell line NS0-derived recombinant human MMR/CD206 Leu19-Lys1383 (Thr399Ala) & (Leu407Phe), Accession # P22897 | |
| 25 μg | |
| Primary | |
| Detects human MMR/CD206 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant mouse (rm) MMR or recombinant human Mrc2 is observed. In Western blots, approximately 25% cross-reactivity with rmMMR is observed. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 685645 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CD206, CLEC13D, CLEC13Dmacrophage mannose receptor 1, C-type lectin domain family 13 member D, mannose receptor, C type 1, MMRCD206 antigen, MRC1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4360 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction