missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-9 Antibody, R&D Systems™

Mouse Monoclonal Antibody
£205.00 - £407.00
Specifications
| Antigen | MMP-9 |
|---|---|
| Clone | 4H3R |
| Dilution | Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18765368
![]() |
R&D Systems
MAB911R-SP |
25 μg |
£205.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18722238
![]() |
R&D Systems
MAB911R-100 |
100 μg |
£407.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-9 Monoclonal specifically detects MMP-9 in Human samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| MMP-9 | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Supernatant | |
| Human | |
| 92 kDa gelatinase, 92 kDa type IV collagenase, CLG4B, EC 3.4.24, EC 3.4.24.35, Gelatinase B, GELB, macrophage gelatinase, MANDP2, matrix metallopeptidase 9, matrix metalloproteinase 9, matrix metalloproteinase-9, MMP9, MMP-9, type V collagenase | |
| Chinese hamster ovary cell line CHO-derived recombinant human MMP-9 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human MMP-9 in direct ELISAs and Western blots. |
| 4H3R | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Mouse | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| 4318 | |
| IgG1 | |
| Protein A or G purified from cell culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title