missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MMP-7 ELISA Kit

Product Code. 17165099
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
Förpackningsstorlek:
Each
Pack of 10
Pack of 96
Denna artikel kan inte returneras. Se returpolicy

Produktkod. 17165099

Brand: Invitrogen™ EH328RBX5

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

ELISA

Human MMP-7 quantitates human MMP-7 in supernatant. The assay will exclusively recognize both natural and recombinant human MMP-7.

Proteins of the matrix metalloproteinase (MMP) family are involved in the breakdown of extracellular matrix in normal physiological processes, such as embryonic development, reproduction, and tissue remodeling, as well as in disease processes, such as arthritis and metastasis. Most MMP's are secreted as inactive proproteins which are activated when cleaved by extracellular proteinases. The enzyme encoded by this gene degrades proteoglycans, fibronectin, elastin and casein and differs from most MMP family members in that it lacks a conserved C-terminal protein domain. The enzyme is involved in wound healing, and studies in mice suggest that it regulates the activity of defensins in intestinal mucosa. The gene is part of a cluster of MMP genes which localize to chromosome 11q22. 3.
TRUSTED_SUSTAINABILITY

Specifikationer

Accession Number HGNC:7174; MAT; Matrilysin; matrilysin, uterine; matrin; matrix metallo protease; matrix metallopeptidase 7; matrix metallopeptidase 7 (matrilysin, uterine); matrix metalloproteinase 7; Matrix metalloproteinase 7 (matrilysin); matrix metalloproteinase 7 (matrilysin, uterine); matrix metalloproteinase-7; MMP; MMP 7; Mmp7; MMP-7; MMP-7 protein; MMPs; MPMM; MPSL1; PUMP 1; Pump 1 protease; PUMP1; PUMP-1; Pump-1 protease; uterine matrilysin; uterine metalloproteinase
Assay Range 0.15 to 100 ng/mL
Assay Sensitivity 0.15 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4316
Gene Symbol MMP7
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 5 x 96 Tests
Regulatory Status RUO
Sample Volume Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target MMP7
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Visa mer Visa mindre
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.