missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-7 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 4 publications
Brand: R&D Systems AF907
This item is not returnable.
View return policy
Description
MMP-7 Polyclonal specifically detects MMP-7 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation, Neutralization.
Specifications
| MMP-7 | |
| Polyclonal | |
| Unconjugated | |
| P09237 | |
| MMP7 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-7 Leu18-Lys267 Accession # P09237 | |
| 100 μg | |
| Primary | |
| Detects human MMP-7 in direct ELISAs and Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse MMP-7 is observed, and less than 1% cross-reactivity with recombinant human (rh) MMP-8 and rhMMP-12 is observed. | |
| Human | |
| IgG |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Neutralization | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 2 ug/mL | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 4316 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction