missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MMP-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 13 publications
£184.00 - £1780.00
Specifications
| Antigen | MMP-7 |
|---|---|
| Clone | 111433 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18750549
![]() |
R&D Systems
MAB9071-SP |
25 μg |
£184.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18777489
![]() |
R&D Systems
MAB9071-100 |
100 μg |
£452.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18749957
![]() |
R&D Systems
MAB9071-500 |
500 μg |
£1780.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-7 Monoclonal specifically detects MMP-7 in Human samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.Specifications
| MMP-7 | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Mouse | |
| Human | |
| NP_002414.1 | |
| 4316 | |
| Mouse myeloma cell line NS0-derived recombinant human MMP-7 Leu18-Lys267 (Ala230del) Accession # NP_002414.1 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects the pro and active forms of human MMP-7 in Western blots. In dot blots, approximately 20% cross-reactivity with recombinant human MMP-8 is observed and no cross-reactivity with recombinant human MMP-1, -2, -3, -9, -10, -12, or -13 is observed. |
| 111433 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, Matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| MMP7 | |
| IgG2b | |
| Protein A or G purified from ascites | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title