missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human MMP-3 Recombinant Protein, PeproTech®

Product Code. 17833883
Click to view available options
Quantity:
1 mg
10 μg
100 μg
2 μg
250 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
250µg
2µg
500µg
50µg
This item is not returnable. View return policy

Product Code. 17833883

Brand: Gibco™ 42003100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Recombinant Human MMP-3 is a 42.8 kDa protein containing the entire catalytic N-terminal domain and the C-terminal domain (378 amino acids). This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08254
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 4314
Molecular Weight (g/mol) 42.8 kDa
Name Human MMP-3
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias CHDS6; EC 3.4.24.17; EMS-2; matrix metallo protease; matrix metallopeptidase 3; matrix metallopeptidase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3; matrix metalloproteinase 3 (stromelysin 1, progelatinase); matrix metalloproteinase 3 precursor; matrix metalloproteinase-3; metalloproteinase 3 receptor; MMP; MMP10; MMP3; MMP-3; MMP-3 protein; MMPs; progelatinase; proteoglycanase; SL-1; SLN1; SLN-1; STMY; STMY1; Str1; STR-1; stromelysin; stromelysin 1; Stromelysin1; stromelysin-1; Transin1; Transin-1
Common Name MMP3
Gene Symbol MMP3
Biological Activity MMP-3 activity was measured by its ability to cleave a chromogenic peptide MMP-3 substrate at room temperature. At an MMP-3 concentration of 2.5 ug/ml, 50% cleavage was achieved (at an incubation time of approximately 75 minutes.)
Conjugate Unconjugated
Recombinant Recombinant
Sequence MRTFPGIPKW RKTHLTYRIV NYTPDLPKDA VDSAVEKALK VWEEVTPLTF SRLYEGEADI MISFAVREHG DFYPFDGPGN VLAHAYAPGP GINGDAHFDD DEQWTKDTTG TNLFLVAAHE IGHSLGLFHS ANTEALMYPL YHSLTDLTRF RLSQDDINGI QSLYGPPPDS PETPLVPTEP VPPEPGTPAN CDPALSFDAV STLRGEILIF KDRHFWRKSL RKLEPELHLI SSFWPSLPSG VDAAYEVTSK DLVFIFKGNQ FWAIRGNEVR AGYPRGIHTL GFPPTVRKID AAISDKEKNK TYFFVEDKYW RFDEKRNSME PGFPKQIAED FPGIDSKIDA VFEEFGFFYF FTGSSQLEFD PNAKKVTHTL KSNSWLNC
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.