missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MMP3 Human ELISA Kit

Product Code. p-7092150
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
96 tests
Pack of 10
This item is not returnable. View return policy

Product Code. 15551487

Brand: Invitrogen™ BMS20143

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-3

For the quantitative detection of total human MMP-3.

MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P08254
Assay Range 0.06-4.00 ng/mL
Assay Sensitivity 0.0005 ng/mL
Conjugate Biotin
Product Type ELISA Kit
Sample Type Plasma, Serum, Supernatant
Target Species Human
For Use With (Application) ELISA
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 4314
Gene Symbol CHDS6,MMP-3,SL-1,STMY,STMY1,STR1
Interassay CV 4%
Intraassay CV 6.1%
Kit Contents Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma, 10 μL; Serum, 10 μL; Supernatant, 10 μL
Storage Requirements 2°C to 8°C
Synonym Matrix metalloproteinase-3, Stromelysin-1, Progelatinase, Transin
Target Human
Test Time 1 hr. 20 min.
Total Assay Time 3 hr. 30 min.
Show More Show Less

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.