missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MMP3 Human ELISA Kit
Description
Includes
Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human MMP-3
MMP3 (stromelysin-1) belongs to the peptidase M10A subfamily. It is secreted in its pro-protein form, and is activated by extracellular proteinase cleavage. MMP3 is capable of binding to zinc and calcium ions, and has metallo-endopeptidase activity. The active enzyme degrades collagen, gelatin, fibronectin, and laminin. It is involved in normal cellular processes such as wound repair and tissue remodeling. Mutations in the gene can result in coronary heart disease 6.
Specifications
Specifications
| Accession Number | P08254 |
| Assay Range | 0.06-4.00 ng/mL |
| Assay Sensitivity | 0.0005 ng/mL |
| Conjugate | Biotin |
| Product Type | ELISA Kit |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Application) | ELISA |
| For Use With (Equipment) | Colorimetric Microplate Reader |
| Gene ID (Entrez) | 4314 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?